siRNA Details
| Gene Symbol: Recql5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130454.2 | NP_569721.1 | 5 | 1518 | ||
| GenBank mRNA | AB038562.1 | - | 3 | 415 | ||
| AB059999.1 | - | 5 | 1032 | |||
| AK029060.1 | - | 5 | 1172 | |||
| AK050965.1 | - | 3 | 247 | |||
| AK083513.1 | - | 5 | 1514 | |||
| BC023769.1 | - | 4 | 810 | |||
| BC053086.1 | - | 5 | 1771 | |||
| BC059000.1 | - | 5 | 1492 | |||
| BC060070.1 | - | 5 | 1771 | |||
| BC066176.1 | - | 5 | 1241 | |||

Recql5
Genomic Map