siRNA Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170985.1 | NP_001164456.1 | 14 | 2130 | ||
| NM_130867.3 | NP_570937.2 | 14 | 2127 | |||
| GenBank mRNA | AF480411.1 | - | 13 | 1765 | ||
| AK158381.1 | - | 14 | 2083 | |||
| AY243095.1 | - | 13 | 1765 | |||
| BC023765.1 | - | 14 | 1943 | 5343720 | ||

Kirrel
Genomic Map