siRNA Details
| Gene Symbol: Krtap21-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028621.3 | NP_082897.2 | 1 | 102 | ||
| GenBank mRNA | AF345297.1 | - | 1 | 96 | ||
| AK003736.2 | - | 1 | 133 | |||
| BC145974.1 | - | 1 | 102 | |||
| BC145976.1 | - | 1 | 102 | |||
| BC147776.1 | - | 1 | 196 | 9087497 | ||
| BC147786.1 | - | 1 | 196 | 9087505 | ||

Krtap21-1
Genomic Map