siRNA Details
| Gene Symbol: Tmem37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019432.2 | NP_062305.1 | 2 | 224 | ||
| GenBank mRNA | AJ272046.1 | - | 2 | 176 | ||
| AK002424.1 | - | 2 | 243 | |||
| AK050134.1 | - | 2 | 224 | |||
| AK086093.1 | - | 2 | 229 | |||
| BC024613.1 | - | 2 | 220 | 4215775 | ||

Tmem37
Genomic Map