siRNA Details
| Gene Symbol: Klra17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133203.5 | NP_573466.3 | 6 | 743 | ||
| GenBank mRNA | AB033769.1 | - | 6 | 756 | ||
| AB190779.1 | - | 5 | 602 | |||
| AB190780.1 | - | 5 | 602 | |||
| AB190781.1 | - | 5 | 602 | |||
| AB190782.1 | - | 5 | 602 | |||
| AB190783.1 | - | 5 | 602 | |||
| AB190784.1 | - | 5 | 602 | |||
| AB190785.1 | - | 5 | 602 | |||
| AB190786.1 | - | 5 | 602 | |||
| AB190787.1 | - | 5 | 602 | |||
| AB190788.1 | - | 5 | 602 | |||
| AB190789.1 | - | 5 | 602 | |||
| AB193831.1 | - | 4 | 459 | |||
| AB193832.1 | - | 6 | 682 | |||
| AB193833.1 | - | 5 | 602 | |||
| AK080021.1 | - | 6 | 744 | |||
| AY620246.1 | - | 5 | 606 | |||
| AY642595.1 | - | 5 | 606 | |||
| BC132236.1 | - | 6 | 704 | 40130513 | ||

Klra17
Genomic Map