siRNA Details
| Gene Symbol: Klra17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133203.5 | NP_573466.3 | 7 | 897 | ||
| GenBank mRNA | AB033769.1 | - | 7 | 910 | ||
| AB190779.1 | - | 6 | 756 | |||
| AB190787.1 | - | 6 | 756 | |||
| AB190788.1 | - | 6 | 756 | |||
| AB190789.1 | - | 6 | 756 | |||
| AB193832.1 | - | 8 | 896 | |||
| AK080021.1 | - | 7 | 898 | |||
| AY642595.1 | - | 6 | 760 | |||
| BC132236.1 | - | 7 | 858 | 40130513 | ||

Klra17
Genomic Map