siRNA Details
| Gene Symbol: Znrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168621.1 | NP_001162092.1 | 2, 3 | 1023 | ||
| NM_001168623.1 | NP_001162094.1 | 2, 3 | 222 | |||
| NM_133206.3 | NP_573469.1 | 2, 3 | 1023 | |||
| GenBank mRNA | AF378525.1 | - | 2, 3 | 728 | ||
| AK030575.1 | - | 1, 2 | 73 | |||
| AK076756.1 | - | 2, 3 | 170 | |||
| AK078819.1 | - | 2, 3 | 249 | |||
| AK079338.1 | - | 2, 3 | 1023 | |||
| AK216619.1 | - | 2, 3 | 149 | |||
| BC006765.1 | - | 2, 3 | 623 | 3710377 | ||
| BC086770.1 | - | 2, 3 | 821 | 5720382 | ||

Znrf1
Genomic Map