siRNA Details
| Gene Symbol: Mtmr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133215.1 | NP_573478.1 | 16 | 2668 | ||
| GenBank mRNA | AF262986.1 | - | 16 | 2668 | ||
| AK146641.1 | - | 16 | 2685 | |||
| BC058091.1 | - | 15 | 2495 | 6822365 | ||
| BC058364.1 | - | 15 | 2495 | 6820754 | ||

Mtmr4
Genomic Map