siRNA Details
Gene Symbol: Zfp704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133218.2 | NP_573481.1 | 2 | 1018 | ||
GenBank mRNA | DT908776.1 | - | 4 | 382 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp704 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133218.2 | NP_573481.1 | 2 | 1018 | ||
GenBank mRNA | DT908776.1 | - | 4 | 382 |