siRNA Details
| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130905.2 | NP_570975.1 | 5, 6 | 500 | ||
| GenBank mRNA | AF373412.1 | - | 5, 6 | 500 | ||
| AK040917.1 | - | 5, 6 | 522 | |||
| BC107188.2 | - | 5, 6 | 500 | 40050810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd209e| Gene Symbol: Cd209e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130905.2 | NP_570975.1 | 5, 6 | 500 | ||
| GenBank mRNA | AF373412.1 | - | 5, 6 | 500 | ||
| AK040917.1 | - | 5, 6 | 522 | |||
| BC107188.2 | - | 5, 6 | 500 | 40050810 | ||