siRNA Details
| Gene Symbol: Cd209a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133238.5 | NP_573501.1 | 3 | 259 | ||
| GenBank mRNA | AF373408.1 | - | 3 | 206 | ||
| AF374470.1 | - | 3 | 206 | |||
| AK054139.1 | - | 3 | 223 | |||
| AK171639.1 | - | 3 | 203 | |||
| AY049062.1 | - | 3 | 259 | |||
| BC111095.1 | - | 2 | 307 | 40040036 | ||

Cd209a
Genomic Map