siRNA Details
| Gene Symbol: Crb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133239.2 | NP_573502.2 | 10 | 3937 | ||
| GenBank mRNA | AF406641.1 | - | 10 | 3902 | ||
| AF492496.1 | - | 9 | 3778 | |||
| AK033059.1 | - | 2 | 50 | |||
| AK164413.1 | - | 2 | 901 | |||
| BC031378.1 | - | 3 | 997 | |||
| BC053331.1 | - | 5 | 2180 | |||

Crb1
Genomic Map