siRNA Details
| Gene Symbol: Il1rl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025602.3 | NP_001020773.1 | 7 | 1068 | ||
| NM_001294171.1 | NP_001281100.1 | 7 | 901 | |||
| NM_010743.3 | NP_034873.2 | 7 | 1068 | |||
| GenBank mRNA | AK033417.1 | - | 7 | 921 | ||
| AK145118.1 | - | 7 | 922 | |||
| BC051445.1 | - | 7 | 878 | 30073421 | ||
| D13695.1 | - | 6 | 760 | |||
| M24843.1 | - | 7 | 909 | |||
| Y07519.1 | - | 7 | 912 | |||

Il1rl1
Genomic Map