siRNA Details
| Gene Symbol: Zfp369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178364.5 | NP_848141.3 | 8 | 1248 | ||
| GenBank mRNA | AJ319726.1 | - | 8 | 1265 | ||
| AK084309.1 | - | 8 | 1251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp369| Gene Symbol: Zfp369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178364.5 | NP_848141.3 | 8 | 1248 | ||
| GenBank mRNA | AJ319726.1 | - | 8 | 1265 | ||
| AK084309.1 | - | 8 | 1251 | |||