siRNA Details
| Gene Symbol: Mcoln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134160.1 | NP_598921.1 | 8 | 987 | ||
| GenBank mRNA | AF475086.1 | - | 8 | 987 | ||
| AK030819.1 | - | 8 | 1107 | |||
| AK033008.1 | - | 8 | 1101 | |||
| AK035029.1 | - | 8 | 1108 | |||
| AY083531.1 | - | 8 | 985 | |||
| BC106856.2 | - | 8 | 987 | 40047039 | ||
| BC106857.2 | - | 8 | 987 | 40047042 | ||

Mcoln3
Genomic Map