siRNA Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134163.4 | NP_598924.1 | 5 | 679 | ||
| GenBank mRNA | AB077700.1 | - | 4 | 585 | ||
| AF466292.1 | - | 4 | 585 | |||
| AJ427920.1 | - | 4 | 585 | |||
| AK040522.1 | - | 4 | 433 | |||
| AK050908.1 | - | 4 | 398 | |||
| AK088971.1 | - | 5 | 692 | |||
| AK167342.1 | - | 4 | 398 | |||
| BC057157.1 | - | 5 | 917 | 5348959 | ||

Mbnl3
Genomic Map