siRNA Details
| Gene Symbol: Ntng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB052336.1 | - | 2 | 241 | ||
| AB052337.1 | - | 2 | 241 | |||
| AB052338.1 | - | 2 | 241 | |||
| AF475079.1 | - | 2 | 241 | |||
| AF475080.1 | - | 2 | 241 | |||
| BC141258.1 | - | 2 | 300 | 9055870 | ||
| BC145600.1 | - | 2 | 300 | 9054166 | ||

Ntng2
Genomic Map