siRNA Details
| Gene Symbol: Galnt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134189.2 | NP_598950.2 | 12 | 1873 | ||
| GenBank mRNA | AK033515.1 | - | 11 | 1531 | ||
| AK131155.1 | - | 12 | 1851 | |||
| AK143714.1 | - | 11 | 1704 | |||
| AK159088.1 | - | 12 | 1872 | |||
| AK159362.1 | - | 12 | 1871 | |||
| AK162424.1 | - | 12 | 1874 | |||
| BC016585.1 | - | 7 | 985 | 3595220 | ||
| BC060617.1 | - | 12 | 1856 | 6836791 | ||

Galnt10
Genomic Map