siRNA Details
| Gene Symbol: Mxd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010753.2 | NP_034883.2 | 5 | 482 | ||
| GenBank mRNA | AK131132.1 | - | 5 | 452 | ||
| AK134169.1 | - | 4 | 463 | |||
| BC011303.1 | - | 5 | 482 | 3490469 | ||
| U32395.1 | - | 5 | 345 | |||

Mxd4
Genomic Map