siRNA Details
| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008539.3 | NP_032565.2 | 5 | 1283 | ||
| GenBank mRNA | AK017583.1 | - | 5 | 1244 | ||
| AK054104.1 | - | 5 | 1244 | |||
| AK084505.1 | - | 5 | 1351 | |||
| AK158509.1 | - | 5 | 1299 | |||
| BC058693.1 | - | 5 | 1283 | 6811514 | ||
| U58992.1 | - | 4 | 956 | |||
| U74359.1 | - | 5 | 1149 | |||

Smad1
Genomic Map