siRNA Details
| Gene Symbol: Smad3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016769.4 | NP_058049.3 | 7 | 1241 | ||
| GenBank mRNA | AB008192.1 | - | 7 | 950 | ||
| AF016189.1 | - | 7 | 1217 | |||
| AK041743.1 | - | 7 | 762 | |||
| AK048626.1 | - | 7 | 1241 | |||
| AK083158.1 | - | 7 | 849 | |||
| BC066850.1 | - | 7 | 927 | 30432720 | ||

Smad3
Genomic Map