siRNA Details
| Gene Symbol: Havcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166631.1 | NP_001160103.1 | 3 | 468 | ||
| NM_001166632.1 | NP_001160104.1 | 3 | 352 | |||
| NM_134248.2 | NP_599009.2 | 2 | 307 | |||
| GenBank mRNA | AF399829.1 | - | 2 | 298 | ||
| AF399830.1 | - | 2 | 298 | |||
| AK133466.1 | - | 3 | 470 | |||
| BC053400.1 | - | 3 | 360 | 30033601 | ||
| CA551363.1 | - | 3 | 352 | |||

Havcr1
Genomic Map