siRNA Details
| Gene Symbol: Creld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133930.1 | NP_598691.1 | 2 | 572 | ||
| GenBank mRNA | AB009020.1 | - | 2 | 577 | ||
| AK042344.1 | - | 2 | 719 | |||
| AK077726.1 | - | 2 | 716 | |||
| AK155777.1 | - | 2 | 582 | |||
| BC010804.1 | - | 2 | 572 | 4239786 | ||
| BC023893.1 | - | 2 | 583 | 5310111 | ||
| BC025932.1 | - | 2 | 478 | 5102571 | ||
| BC029065.1 | - | 2 | 608 | 4935296 | ||

Creld1
Genomic Map