siRNA Details
| Gene Symbol: Mfap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161799.1 | NP_001155271.1 | 9 | 559 | ||
| NM_008546.3 | NP_032572.2 | 9 | 585 | |||
| GenBank mRNA | AK160213.1 | - | 9 | 561 | ||
| AK164920.1 | - | 9 | 542 | |||
| BC035490.1 | - | 9 | 519 | 5249342 | ||
| DV654229.1 | - | 9 | 559 | |||
| L23769.1 | - | 9 | 541 | |||

Mfap2
Genomic Map