siRNA Details
| Gene Symbol: Mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010762.5 | NP_034892.1 | 2 | 782 | ||
| GenBank mRNA | AK004639.1 | - | 2 | 369 | ||
| AK158653.1 | - | 2 | 783 | |||
| AK169172.1 | - | 2 | 306 | |||
| BC006826.1 | - | 2 | 279 | 3597561 | ||
| Y07626.1 | - | 2 | 258 | |||
| Y07812.1 | - | 2 | 220 | |||

Mal
Genomic Map