siRNA Details
Gene Symbol: Fxyd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008557.2 | NP_032583.1 | 2, 3 | 139 | ||
GenBank mRNA | AA959274.1 | - | 2, 3 | 96 | ||
AK145206.1 | - | 110 | ||||
AK166426.1 | - | 2, 3 | 112 | |||
BC002039.1 | - | 2, 3 | 103 | 3488180 | ||
BC051033.1 | - | 2, 3 | 97 | 4460667 | ||
BC056223.1 | - | 2, 3 | 107 | 6485767 | ||
BG067532.2 | - | 6, 7 | 382 | |||
X93038.1 | - | 2, 3 | 135 |
