siRNA Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072248.1 | - | 4, 5 | 758 | ||
| AK185793.1 | - | 1, 2 | 80 | |||
| BC038264.1 | - | 4, 5 | 559 | 5250237 | ||
| BQ042794.1 | - | 3, 4 | 326 | |||
| CK625083.1 | - | 4, 5 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mbd3| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF072248.1 | - | 4, 5 | 758 | ||
| AK185793.1 | - | 1, 2 | 80 | |||
| BC038264.1 | - | 4, 5 | 559 | 5250237 | ||
| BQ042794.1 | - | 3, 4 | 326 | |||
| CK625083.1 | - | 4, 5 | 578 | |||