siRNA Details
| Gene Symbol: Mc1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008559.2 | NP_032585.2 | 1 | 575 | ||
| GenBank mRNA | AK148437.1 | - | 1 | 551 | ||
| BC119294.1 | - | 1 | 249 | 8734043 | ||
| BC119296.1 | - | 1 | 249 | 8734045 | ||
| BY740794.1 | - | 1 | 575 | |||
| X65635.1 | - | 1 | 161 | |||
More Information
Set Membership: |
|

Mc1r
Genomic Map