siRNA Details
| Gene Symbol: Mcf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159485.1 | NP_001152957.1 | 24 | 2904 | ||
| NM_001159486.1 | NP_001152958.1 | 24 | 2904 | |||
| NM_178076.3 | NP_835177.2 | 24 | 2807 | |||
| GenBank mRNA | AK122259.1 | - | 15 | 1712 | ||
| AK147252.1 | - | 21 | 3011 | |||
| AK170186.1 | - | 24 | 2865 | |||
| AK170558.1 | - | 24 | 2838 | |||
| BC029315.1 | - | 5 | 490 | |||
| BC058622.1 | - | 24 | 2742 | 5704822 | ||
| CF618259.1 | - | 1 | 23 | |||
| DQ978380.1 | - | 23 | 2658 | |||
| S76838.1 | - | 25 | 3101 | |||

Mcf2l
Genomic Map