siRNA Details
| Gene Symbol: Mcpt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010779.2 | NP_034909.2 | 4 | 405 | ||
| GenBank mRNA | AB051900.1 | - | 4 | 353 | ||
| AK144385.1 | - | 4 | 406 | |||
| AY007569.1 | - | 4 | 405 | |||
| BC026198.1 | - | 4 | 381 | 1314753 | ||
| CJ046459.1 | - | 4 | 408 | |||
| M55617.1 | - | 405 | ||||
| X68804.1 | - | 4 | 352 | |||

Mcpt4
Genomic Map