siRNA Details
| Gene Symbol: Smcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008574.3 | NP_032600.2 | 1 | 92 | ||
| GenBank mRNA | AK077172.1 | - | 1 | 63 | ||
| BC055104.1 | - | 1 | 62 | 6773089 | ||
| BC061167.1 | - | 1 | 42 | 6771333 | ||
| BU936528.1 | - | 1 | 95 | |||
| M29603.1 | - | 1 | 65 | |||
| S49659.1 | - | 1 | 62 | |||
| S49660.1 | - | 212 | ||||

Smcp
Genomic Map