siRNA Details
| Gene Symbol: Meis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193271.1 | NP_001180200.1 | 7 | 1379 | ||
| NM_010789.3 | NP_034919.1 | 7 | 1379 | |||
| GenBank mRNA | AB221660.1 | - | 6 | 642 | ||
| AK049720.1 | - | 6 | 1191 | |||
| AK132298.1 | - | 7 | 1159 | |||
| AK134321.1 | - | 7 | 1374 | |||
| AK140748.1 | - | 7 | 1383 | |||
| AK146680.1 | - | 3 | 187 | |||
| AK168684.1 | - | 7 | 1371 | |||
| BC023689.1 | - | 7 | 1375 | 5344307 | ||
| U33629.1 | - | 7 | 1090 | |||
| U33630.1 | - | 7 | 1090 | |||

Meis1
Genomic Map