siRNA Details
| Gene Symbol: Meox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010791.3 | NP_034921.1 | 1 | 505 | ||
| GenBank mRNA | AK052370.1 | - | 1 | 515 | ||
| BC011082.1 | - | 1 | 505 | 3984366 | ||
| Z15103.1 | - | 1 | 460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Meox1| Gene Symbol: Meox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010791.3 | NP_034921.1 | 1 | 505 | ||
| GenBank mRNA | AK052370.1 | - | 1 | 515 | ||
| BC011082.1 | - | 1 | 505 | 3984366 | ||
| Z15103.1 | - | 1 | 460 | |||