siRNA Details
| Gene Symbol: Mertk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008587.1 | NP_032613.1 | 19 | 2637 | ||
| GenBank mRNA | AK029009.1 | - | 8 | 839 | ||
| AK030158.1 | - | 11 | 1336 | |||
| AK079252.1 | - | 2 | 250 | |||
| L11625.1 | - | 10 | 1175 | |||
| U21301.1 | - | 19 | 2637 | |||

Mertk
Genomic Map