siRNA Details
| Gene Symbol: Mesp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008588.2 | NP_032614.2 | 1 | 146 | ||
| GenBank mRNA | BC012689.1 | - | 1 | 355 | ||
| BC125505.1 | - | 1 | 146 | 40130020 | ||
| BC125507.1 | - | 1 | 146 | 40130022 | ||
| D83674.1 | - | 1 | 209 | |||

Mesp1
Genomic Map