siRNA Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008591.2 | NP_032617.2 | 22 | 4414 | ||
| GenBank mRNA | BC117969.1 | - | 20 | 4058 | ||
| BC117970.1 | - | 20 | 4054 | |||
| BC128481.1 | - | 1 | 127 | |||
| BC128482.1 | - | 2 | 125 | |||
| CX563675.1 | - | 3 | 372 | |||
| Y00671.1 | - | 20 | 4059 | |||

Met
Genomic Map