siRNA Details
Gene Symbol: Foxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008593.3 | NP_032619.1 | 1 | 459 | ||
GenBank mRNA | AF023915.1 | - | 1 | 437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Foxd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008593.3 | NP_032619.1 | 1 | 459 | ||
GenBank mRNA | AF023915.1 | - | 1 | 437 |