siRNA Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008598.2 | NP_032624.1 | 2 | 178 | ||
GenBank mRNA | AB092489.1 | - | 1 | 89 | ||
AB092490.1 | - | 1 | 89 | |||
AK090389.1 | - | 167 | ||||
BC031888.1 | - | 2 | 147 | 5372377 | ||
BC096557.1 | - | 2 | 181 | 1366095 | ||
BY344992.1 | - | 2 | 186 | |||
M84524.1 | - | 1 | 104 |
