siRNA Details
Gene Symbol: Pias2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164167.1 | NP_001157639.1 | 12 | 1710 | ||
NM_001164168.1 | NP_001157640.1 | 12 | 1726 | |||
NM_001164169.1 | NP_001157641.1 | 12 | 1710 | |||
NM_001164170.1 | NP_001157642.1 | 12 | 1726 | |||
NM_008602.4 | NP_032628.3 | 12 | 1710 | |||
GenBank mRNA | AF039567.1 | - | 11 | 1155 | ||
AF201391.1 | - | 12 | 1637 | |||
AK014871.1 | - | 12 | 1701 | |||
AK029716.1 | - | 12 | 1711 | |||
AK032463.1 | - | 2 | 145 | |||
AK078097.1 | - | 7 | 777 | |||
AK078813.1 | - | 12 | 1680 | |||
AK086653.1 | - | 12 | 1642 | |||
AK149142.1 | - | 13 | 1749 | |||
BC005596.1 | - | 12 | 1587 | 3969546 | ||
BC034711.1 | - | 12 | 1716 | 2631738 |
