siRNA Details
| Gene Symbol: Mllt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252560.1 | NP_001239489.1 | 6 | 740 | ||
| NM_001252561.1 | NP_001239490.1 | 7 | 791 | |||
| NM_010804.4 | NP_034934.2 | 6 | 595 | |||
| GenBank mRNA | AF010135.1 | - | 6 | 638 | ||
| AK045042.1 | - | 6 | 495 | |||
| AK147388.1 | - | 9 | 1085 | |||
| AK154320.1 | - | 6 | 740 | |||
| AK158426.1 | - | 5 | 333 | |||
| AK220504.1 | - | 8 | 847 | |||
| BC055298.1 | - | 7 | 740 | |||
| BC069988.1 | - | 5 | 589 | |||
| BC070475.1 | - | 6 | 561 | 30638433 | ||
| BY608946.1 | - | 1 | 39 | |||
| CN534499.1 | - | 7 | 628 | |||
| CN537566.1 | - | 3 | 268 | |||

Mllt10
Genomic Map