siRNA Details
| Gene Symbol: Aff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080798.1 | NP_001074267.1 | 14 | 3009 | ||
| NM_133919.3 | NP_598680.3 | 14 | 3020 | |||
| GenBank mRNA | AF013131.1 | - | 13 | 2835 | ||
| AF074266.1 | - | 14 | 2850 | |||
| AY749168.1 | - | 14 | 2853 | |||
| BC131665.1 | - | 14 | 2853 | 40110795 | ||
| CN537099.1 | - | 1 | 18 | |||

Aff1
Genomic Map