siRNA Details
Gene Symbol: Trpm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039104.2 | NP_001034193.2 | 14 | 2005 | ||
NM_018752.3 | NP_061222.3 | 14 | 1898 | |||
GenBank mRNA | AF047714.1 | - | 14 | 1767 | ||
AK044507.1 | - | 14 | 1929 | |||
AK148193.1 | - | 12 | 1847 | |||
AK148268.1 | - | 14 | 1752 | |||
AK148275.1 | - | 14 | 1808 | |||
AK149290.1 | - | 14 | 1927 | |||
AK181524.1 | - | 11 | ||||
AK191136.1 | - | 11 | ||||
AK191215.1 | - | 11 | ||||
AK196486.1 | - | 4 | 326 | |||
AK197773.1 | - | 11 | ||||
AK198555.1 | - | 11 | ||||
AK198807.1 | - | 11 | ||||
AK199374.1 | - | 11 | ||||
AK199794.1 | - | 11 | ||||
AK203767.1 | - | 11 | ||||
AK204139.1 | - | 11 | ||||
AK205296.1 | - | 11 | ||||
AK208211.1 | - | 1 | 38 | |||
AK209819.1 | - | 11 | ||||
AY180104.2 | - | 14 | 2005 |
