siRNA Details
| Gene Symbol: Mpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122949.2 | NP_001116421.1 | 9, 10 | 1511 | ||
| NM_001285496.1 | NP_001272425.1 | 9, 10 | 1331 | |||
| NM_010823.3 | NP_034953.2 | 9, 10 | 1487 | |||
| GenBank mRNA | AF360122.1 | - | 9, 10 | 1268 | ||
| AK041166.1 | - | 9, 10 | 1459 | |||
| BC103513.1 | - | 9, 10 | 1444 | |||
| BC103515.1 | - | 9, 10 | 1468 | |||
| BC146612.1 | - | 9, 10 | 1440 | |||
| BC146613.1 | - | 9, 10 | 1445 | 40039147 | ||
| X73677.1 | - | 9, 10 | 1455 | |||
| Z22649.1 | - | 9, 10 | 1445 | |||

Mpl
Genomic Map