siRNA Details
Gene Symbol: Mstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010834.2 | NP_034964.1 | 2 | 709 | ||
GenBank mRNA | AK132415.1 | - | 2 | 734 | ||
AK142222.1 | - | 2 | 732 | |||
BC103676.1 | - | 2 | 709 | 40047210 | ||
BC103677.1 | - | 2 | 709 | 40047209 | ||
BC103678.1 | - | 2 | 709 | 40047208 | ||
BC105674.1 | - | 2 | 709 | 40047211 | ||
U84005.1 | - | 2 | 709 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
