siRNA Details
| Gene Symbol: Msx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010836.3 | NP_034966.1 | 2 | 434 | ||
| GenBank mRNA | AB221640.1 | - | 2 | 345 | ||
| AK012556.1 | - | 2 | 503 | |||
| BC051983.1 | - | 2 | 403 | |||
| BC137577.1 | - | 2 | 433 | 8860596 | ||
| BC137578.1 | - | 2 | 433 | 8860597 | ||
| U62523.1 | - | 2 | 441 | |||

Msx3
Genomic Map