siRNA Details
| Gene Symbol: Map7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198635.1 | NP_001185564.1 | 9 | 1061 | ||
| NM_008635.2 | NP_032661.2 | 9 | 1037 | |||
| GenBank mRNA | AK014575.1 | - | 9 | 954 | ||
| AK133267.1 | - | 9 | 1486 | |||
| BC042771.1 | - | 9 | 962 | |||
| BC044733.1 | - | 9 | 962 | |||
| BC046555.1 | - | 9 | 962 | |||
| BC052637.1 | - | 9 | 997 | 6467153 | ||

Map7
Genomic Map