siRNA Details
Gene Symbol: Mapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038609.2 | NP_001033698.1 | 11 | 1213 | ||
NM_001285454.1 | NP_001272383.1 | 8 | 946 | |||
NM_001285455.1 | NP_001272384.1 | 8 | 973 | |||
NM_001285456.1 | NP_001272385.1 | 8 | 946 | |||
NM_010838.4 | NP_034968.3 | 9 | 1039 | |||
GenBank mRNA | AF483518.1 | - | 8 | 854 | ||
AF483519.1 | - | 8 | 854 | |||
AK078167.1 | - | 9 | 1011 | |||
AK147636.1 | - | 11 | 1174 | |||
BC014748.1 | - | 9 | 977 | 4504850 | ||
M18775.1 | - | 8 | 947 | |||
M18776.1 | - | 8 | 947 | |||
M93266.1 | - | 4 | 358 | |||
U12914.1 | - | 11 | 1097 | |||
U12915.1 | - | 9 | 923 | |||
U12916.1 | - | 8 | 857 | |||
Z12133.1 | - | 4 | 358 |
