siRNA Details
| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023166.2 | NP_075655.2 | 2 | 644 | ||
| NM_181452.2 | NP_852117.2 | 2 | 683 | |||
| GenBank mRNA | AB012726.1 | - | 2 | 668 | ||
| AF212920.1 | - | 2 | 665 | |||
| AF212921.1 | - | 2 | 611 | |||
| AK003883.1 | - | 2 | 633 | |||
| AK013256.1 | - | 2 | 612 | |||
| BC013894.1 | - | 2 | 671 | 4217979 | ||
| BC056166.1 | - | 2 | 629 | 5715229 | ||
| BM942073.1 | - | 2 | 609 | |||
| CJ132526.1 | - | 2 | 644 | |||

Fam89b
Genomic Map