siRNA Details
| Gene Symbol: Muc5ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010844.1 | NP_034974.1 | 33 | 5774 | ||
| GenBank mRNA | L42292.1 | - | 3 | 1328 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc5ac| Gene Symbol: Muc5ac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010844.1 | NP_034974.1 | 33 | 5774 | ||
| GenBank mRNA | L42292.1 | - | 3 | 1328 | ||