siRNA Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013606.1 | NP_038634.1 | 11, 12 | 1524 | ||
| GenBank mRNA | AB029920.1 | - | 11, 12 | 1524 | ||
| AK143831.1 | - | 11, 12 | 1528 | |||
| AK147949.1 | - | 2147 | ||||
| BC007127.1 | - | 11, 12 | 1509 | 3591798 | ||
| J03368.1 | - | 11, 12 | 1525 | |||

Mx2
Genomic Map